Assay Details
| Gene Symbol: CFAP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003513.4 | XP_004003562.2 | 19 - 20 | 3233 | 54 | |
| XM_012116834.3 | XP_011972224.2 | 18 - 19 | 3020 | 54 | ||
| XM_027968647.1 | XP_027824448.1 | 19 - 20 | 3233 | 54 | ||
More Information
Set Membership: |
|

CFAP57
Genomic Map