Assay Details
Gene Symbol: DEPDC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979605.1 | XP_027835406.1 | 8 - 9 | 1364 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
