Assay Details
| Gene Symbol: EEF1AKNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013696.4 | XP_004013745.2 | 6 - 7 | 2007 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EEF1AKNMT| Gene Symbol: EEF1AKNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013696.4 | XP_004013745.2 | 6 - 7 | 2007 | 102 | |
Set Membership: |
|