Assay Details
Gene Symbol: LOC105602050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977615.1 | XP_027833416.1 | 6 - 7 | 624 | 68 | |
XM_027977616.1 | XP_027833417.1 | 4 - 5 | 497 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
