Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015209.4 | XP_004015258.2 | 1 - 2 | 191 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP8| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015209.4 | XP_004015258.2 | 1 - 2 | 191 | 83 | |