Assay Details
Gene Symbol: LOC114118096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978907.1 | XP_027834708.1 | 2 - 3 | 342 | 73 |
Gene Symbol: LOC101107463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978908.1 | XP_027834709.1 | 2 - 3 | 352 | 73 |
