Assay Details
| Gene Symbol: CARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959343.1 | XP_027815144.1 | 14 - 15 | 1693 | 64 | |
| XM_027959344.1 | XP_027815145.1 | 13 - 14 | 1441 | 64 | ||
More Information
Set Membership: |
|

CARS1
Genomic Map