Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004067.4 | XP_004004116.2 | 8 - 9 | 2362 | 76 | |
| XM_012115851.3 | XP_011971241.1 | 9 - 10 | 2191 | 76 | ||
| XM_027964189.1 | XP_027819990.1 | 9 - 10 | 2484 | 76 | ||
| XM_027964190.1 | XP_027819991.1 | 8 - 9 | 2301 | 76 | ||
More Information
Set Membership: |
|

PTPDC1
Genomic Map