Assay Details
| Gene Symbol: IFNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011371.4 | XP_004011420.3 | 1 - 2 | 278 | 72 | |
| XM_012182970.3 | XP_012038360.2 | 1 - 2 | 214 | 72 | ||
More Information
Set Membership: |
|

IFNGR1
Genomic Map