Assay Details
| Gene Symbol: OSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973182.1 | XP_027828983.1 | 2 - 3 | 1157 | 69 | |
| XM_027973183.1 | XP_027828984.1 | 2 - 3 | 1157 | 69 | ||
More Information
Set Membership: |
|

OSR2
Genomic Map