Assay Details
| Gene Symbol: BACH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015091726.2 | XP_014947212.2 | 2 - 3 | 1790 | 74 | |
| XM_027978489.1 | XP_027834290.1 | 3 - 4 | 1690 | 74 | ||
| XM_027978499.1 | XP_027834300.1 | 3 - 4 | 1769 | 74 | ||
More Information
Set Membership: |
|

BACH1
Genomic Map