Assay Details
| Gene Symbol: LOC101103154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009901.4 | XP_004009950.2 | 5 - 6 | 895 | 67 | |
| XM_027971062.1 | XP_027826863.1 | 5 - 6 | 890 | 67 | ||
More Information
Set Membership: |
|

LOC101103154
Genomic Map