Assay Details
| Gene Symbol: PROX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187757.3 | XP_012043147.2 | 3 - 4 | 2499 | 87 | |
| XM_012187758.3 | XP_012043148.2 | 3 - 4 | 2770 | 87 | ||
| XM_015099366.2 | XP_014954852.2 | 3 - 4 | 2053 | 87 | ||
| XM_015099368.2 | XP_014954854.2 | 4 - 5 | 2188 | 87 | ||
| XM_027975966.1 | XP_027831767.1 | 3 - 4 | 1988 | 87 | ||
| XM_027975967.1 | XP_027831768.1 | 3 - 4 | 3237 | 87 | ||
| XM_027975968.1 | XP_027831769.1 | 3 - 4 | 2770 | 87 | ||
More Information
Set Membership: |
|

PROX1
Genomic Map