Assay Details
| Gene Symbol: KCTD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956644.1 | XP_027812445.1 | 2 - 3 | 287 | 61 | |
| XM_027956645.1 | XP_027812446.1 | 2 - 3 | 287 | 61 | ||
More Information
Set Membership: |
|

KCTD10
Genomic Map