Assay Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958998.1 | XP_027814799.1 | 5 - 6 | 884 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958998.1 | XP_027814799.1 | 5 - 6 | 884 | 61 | |
Set Membership: |
|