Assay Details
| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106019.3 | XP_011961409.1 | 5 - 6 | 1244 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRNB3| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106019.3 | XP_011961409.1 | 5 - 6 | 1244 | 61 | |
Set Membership: |
|