Assay Details
| Gene Symbol: RAD9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959301.1 | XP_027815102.1 | 8 - 9 | 1061 | 56 | |
| XM_027959302.1 | XP_027815103.1 | 9 - 10 | 986 | 56 | ||
More Information
Set Membership: |
|

RAD9A
Genomic Map