Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976754.1 | XP_027832555.1 | 6 - 7 | 853 | 66 | |
| XM_027976757.1 | XP_027832558.1 | 6 - 7 | 853 | 66 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIRREL1| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976754.1 | XP_027832555.1 | 6 - 7 | 853 | 66 | |
| XM_027976757.1 | XP_027832558.1 | 6 - 7 | 853 | 66 | ||