Assay Details
| Gene Symbol: DELE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008905.4 | XP_004008954.2 | 4 - 5 | 515 | 68 | |
| XM_012178656.3 | XP_012034046.2 | 4 - 5 | 515 | 68 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DELE1| Gene Symbol: DELE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008905.4 | XP_004008954.2 | 4 - 5 | 515 | 68 | |
| XM_012178656.3 | XP_012034046.2 | 4 - 5 | 515 | 68 | ||