Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092937.2 | XP_014948423.2 | 2 - 3 | 310 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALKAL2| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092937.2 | XP_014948423.2 | 2 - 3 | 310 | 80 | |