Assay Details
| Gene Symbol: MFSD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011484.4 | XP_004011533.4 | 3 - 4 | 364 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFSD4B| Gene Symbol: MFSD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011484.4 | XP_004011533.4 | 3 - 4 | 364 | 64 | |
Set Membership: |
|