Assay Details
| Gene Symbol: OBI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012192.4 | XP_004012241.2 | 1 - 2 | 116 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OBI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012192.4 | XP_004012241.2 | 1 - 2 | 116 | 93 | |