Assay Details
Gene Symbol: OBI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012192.4 | XP_004012241.2 | 3 - 4 | 338 | 90 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OBI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012192.4 | XP_004012241.2 | 3 - 4 | 338 | 90 |