Assay Details
| Gene Symbol: ACSBG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098628.3 | XP_011954018.2 | 13 - 14 | 2106 | 71 | |
| XM_027957149.1 | XP_027812950.1 | 14 - 15 | 3053 | 71 | ||
More Information
Set Membership: |
|

ACSBG1
Genomic Map