Assay Details
| Gene Symbol: SUPT3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102730.2 | XP_014958216.1 | 10 - 11 | 1163 | 70 | |
| XM_015102732.2 | XP_014958218.1 | 10 - 11 | 1163 | 70 | ||
| XM_015102734.2 | XP_014958220.1 | 10 - 11 | 1250 | 70 | ||
| XM_015102735.2 | XP_014958221.1 | 10 - 11 | 1163 | 70 | ||
| XM_027959126.1 | XP_027814927.1 | 10 - 11 | 1163 | 70 | ||
More Information
Set Membership: |
|

SUPT3H
Genomic Map