Assay Details
| Gene Symbol: LOC101102402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975350.1 | XP_027831151.1 | 18 - 19 | 2069 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101102402| Gene Symbol: LOC101102402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975350.1 | XP_027831151.1 | 18 - 19 | 2069 | 81 | |