Assay Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107412.3 | XP_011962802.1 | 12 - 13 | 1545 | 69 | |
| XM_015105060.2 | XP_014960546.1 | 12 - 13 | 1522 | 69 | ||
| XM_015105061.2 | XP_014960547.1 | 12 - 13 | 1515 | 69 | ||
| XM_015105062.2 | XP_014960548.1 | 11 - 12 | 1351 | 69 | ||
More Information
Set Membership: |
|

ZNF280C
Genomic Map