Assay Details
| Gene Symbol: LOC101102057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008561.4 | XP_004008610.1 | 2 - 3 | 187 | 75 | |
| XM_027969726.1 | XP_027825527.1 | 2 - 3 | 187 | 75 | ||
More Information
Set Membership: |
|

LOC101102057
Genomic Map