Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009036.3 | XP_004009085.2 | 4 - 5 | 454 | 81 | |
| XM_012178886.3 | XP_012034276.2 | 4 - 5 | 429 | 81 | ||
| XM_012178887.2 | XP_012034277.2 | 4 - 5 | 451 | 81 | ||
More Information
Set Membership: |
|

PTTG1
Genomic Map