Assay Details
| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964294.1 | XP_027820095.1 | 4 - 5 | 587 | 72 | |
| GenBank mRNA | AF123316.1 | - | 2 - 3 | 260 | 72 | |
More Information
Set Membership: |
|

AQP3
Genomic Map