Assay Details
| Gene Symbol: VCPKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010506.4 | XP_004010555.1 | 4 - 5 | 1097 | 98 | |
| XM_012181322.3 | XP_012036712.1 | 3 - 4 | 1013 | 98 | ||
More Information
Set Membership: |
|

VCPKMT
Genomic Map