Assay Details
| Gene Symbol: PDIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094221.2 | XP_014949707.1 | 9 - 10 | 1033 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDIA6| Gene Symbol: PDIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094221.2 | XP_014949707.1 | 9 - 10 | 1033 | 73 | |
Set Membership: |
|