Assay Details
| Gene Symbol: LOC101111154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021351.4 | XP_004021400.2 | 3 - 4 | 557 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111154| Gene Symbol: LOC101111154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021351.4 | XP_004021400.2 | 3 - 4 | 557 | 100 | |