Assay Details
| Gene Symbol: CSGALNACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105064.3 | XP_011960454.1 | 3 - 4 | 1259 | 89 | |
| XM_027962336.1 | XP_027818137.1 | 3 - 4 | 1259 | 89 | ||
More Information
Set Membership: |
|

CSGALNACT2
Genomic Map