Assay Details
| Gene Symbol: ITPRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008240.4 | XP_004008289.3 | 6 - 7 | 467 | 72 | |
| XM_027968636.1 | XP_027824437.1 | 5 - 6 | 456 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPRID1| Gene Symbol: ITPRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008240.4 | XP_004008289.3 | 6 - 7 | 467 | 72 | |
| XM_027968636.1 | XP_027824437.1 | 5 - 6 | 456 | 72 | ||