Assay Details
| Gene Symbol: ITPRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008240.4 | XP_004008289.3 | 7 - 8 | 543 | 109 | |
| XM_027968636.1 | XP_027824437.1 | 6 - 7 | 532 | 109 | ||
More Information
Set Membership: |
|

ITPRID1
Genomic Map