Assay Details
| Gene Symbol: LOC101114042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099914.2 | XP_014955400.1 | 2 - 3 | 471 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114042| Gene Symbol: LOC101114042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099914.2 | XP_014955400.1 | 2 - 3 | 471 | 64 | |
Set Membership: |
|