Assay Details
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187925.3 | XP_012043315.3 | 4 - 5 | 768 | 65 | |
XM_015099175.2 | XP_014954661.1 | 4 - 5 | 735 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
