Assay Details
| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099184.2 | XP_014954670.2 | 11 - 12 | 1458 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCCPDH| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099184.2 | XP_014954670.2 | 11 - 12 | 1458 | 60 | |
Set Membership: |
|