Assay Details
| Gene Symbol: HELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104564.2 | XP_014960050.1 | 2 - 3 | 135 | 71 | |
| XM_027962644.1 | XP_027818445.1 | 2 - 3 | 135 | 71 | ||
More Information
Set Membership: |
|

HELT
Genomic Map