Assay Details
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969546.1 | XP_027825347.1 | 2 - 3 | 192 | 68 | |
XM_027969547.1 | XP_027825348.1 | 2 - 3 | 202 | 68 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969546.1 | XP_027825347.1 | 2 - 3 | 192 | 68 | |
XM_027969547.1 | XP_027825348.1 | 2 - 3 | 202 | 68 |