Assay Details
| Gene Symbol: SLC23A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970288.1 | XP_027826089.1 | 13 - 14 | 1828 | 66 | |
| XM_027970289.1 | XP_027826090.1 | 13 - 14 | 1829 | 66 | ||
| XM_027970290.1 | XP_027826091.1 | 13 - 14 | 1830 | 66 | ||
| XM_027970291.1 | XP_027826092.1 | 14 - 15 | 1704 | 66 | ||
| XM_027970292.1 | XP_027826093.1 | 11 - 12 | 1524 | 66 | ||
More Information
Set Membership: |
|

SLC23A1
Genomic Map