Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004166.3 | XP_004004215.1 | 1 - 2 | 913 | 74 | |
| XM_012118802.2 | XP_011974192.1 | 1 - 2 | 913 | 74 | ||
| GenBank mRNA | EU723257.1 | - | 1 - 2 | 298 | 74 | |
More Information
Set Membership: |
|

ADRA1A
Genomic Map