Assay Details
| Gene Symbol: HAUS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180992.2 | XP_012036382.1 | 3 - 4 | 352 | 61 | |
| XM_015096884.2 | XP_014952370.1 | 3 - 4 | 570 | 61 | ||
| XM_027971643.1 | XP_027827444.1 | 3 - 4 | 322 | 61 | ||
| XM_027971644.1 | XP_027827445.1 | 3 - 4 | 534 | 61 | ||
More Information
Set Membership: |
|

HAUS4
Genomic Map