Assay Details
| Gene Symbol: IRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095774.3 | XP_011951164.2 | 12 - 13 | 1628 | 64 | |
| XM_027979395.1 | XP_027835196.1 | 11 - 12 | 1572 | 64 | ||
| XM_027979396.1 | XP_027835197.1 | 12 - 13 | 1625 | 64 | ||
More Information
Set Membership: |
|

IRAG1
Genomic Map