Assay Details
| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101700.3 | XP_011957090.2 | 4 - 5 | 426 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDIAS| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101700.3 | XP_011957090.2 | 4 - 5 | 426 | 75 | |
Set Membership: |
|