Assay Details
| Gene Symbol: LOC101118248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021901.4 | XP_004021950.2 | 4 - 5 | 614 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118248| Gene Symbol: LOC101118248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021901.4 | XP_004021950.2 | 4 - 5 | 614 | 115 | |
Set Membership: |
|