Assay Details
| Gene Symbol: LOC101118248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021901.4 | XP_004021950.2 | 6 - 7 | 815 | 61 | |
| Gene Symbol: LOC114113165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964882.1 | XP_027820683.1 | 1 | 757 | 61 | |
More Information
Set Membership: |
|

LOC101118248
Genomic Map