Assay Details
| Gene Symbol: LOC101122274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096076.2 | XP_014951562.2 | 2 - 3 | 3090 | 76 | |
| XM_027970324.1 | XP_027826125.1 | 2 - 3 | 2993 | 76 | ||
| XM_027970325.1 | XP_027826126.1 | 2 - 3 | 3011 | 76 | ||
More Information
Set Membership: |
|

LOC101122274
Genomic Map