Assay Details
| Gene Symbol: LOC101122274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096076.2 | XP_014951562.2 | 3 - 4 | 3182 | 92 | |
| XM_027970324.1 | XP_027826125.1 | 3 - 4 | 3085 | 92 | ||
| XM_027970325.1 | XP_027826126.1 | 3 - 4 | 3103 | 92 | ||
More Information
Set Membership: |
|

LOC101122274
Genomic Map